Learn More
Aviva Systems Biology Corp TCF12 Antibody - N-terminal region (P100771_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100771_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCF12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TCF12. Alternate names: HEB, HTF4, HsT17266, bHLHb20ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Zebrafish
Catalog No. 50-014-62705