Learn More
Aviva Systems Biology Corp TCF25 Antibody - N-terminal region (ARP47624_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47624_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TCF25. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TCF25. Alternate names: FKSG26, Hulp1, KIAA1049, NULP1, PRO2620, hKIAA1049ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-59613