Learn More
Aviva Systems Biology Corp TCF7 Antibody - middle region (ARP31429_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31429_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCF7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TCF7. Alternate names: MGC47735, TCF-1ApplicationWB. Species reactivity: Human
Catalog No. 50-014-54217