Learn More
Aviva Systems Biology Corp TCF7 Antibody - middle region (ARP34782_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34782_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCF7. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human TCF7. Alternate names: TCF-1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-55561