Learn More
Aviva Systems Biology Corp TCF7L1 Antibody - N-terminal region (ARP57917_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57917_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCF7L1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TCF7L1. Alternate names: TCF-3, TCF3ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53759