Learn More
Aviva Systems Biology Corp TCF7L2 Antibody - N-terminal region (ARP39650_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39650_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tcf7l2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TCF7L2. Alternate names: TCF4, TCF-4ApplicationWB. Species reactivity: Human
Catalog No. 50-284-819