Learn More
Aviva Systems Biology Corp TCFAP2C Antibody - N-terminal region (ARP40141_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40141_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TCFAP2C. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of mouse TCFAP2C. Alternate names: Stra2, Ap-2.2, Tcfap2c, AA409384, AP2gammaApplicationWB. Species reactivity: Mouse, Rat
Catalog No. 50-014-57306