Learn More
Aviva Systems Biology Corp TCFAP4 Antibody - middle region (ARP37416_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37416_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TCFAP4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse TCFAP4. Alternate names: AP-4, Tcfap4, bHLHc41, AI642933, D930048N17RikApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56365