Learn More
Aviva Systems Biology Corp Tcfap4 Antibody - middle region (ARP37415_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37415_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tcfap4. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of mouse Tcfap4. Alternate names: AI642933, AP-4, D930048N17Rik, Tfap4, Tcfap4, bHLHc41ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-629