Learn More
Aviva Systems Biology Corp TCFL5 Antibody - middle region (ARP31500_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31500_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCFL5. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the middle region of human TCFL5. Alternate names: CHA, Figlb, E2BP-1, bHLHe82ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-54246