Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ TCG-ProbeSwabQC™
GSA_VA

Catalog No. R485103 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 Loops/Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R485103 5 Loops/Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R485103 Supplier Thermo Scientific™ Supplier No. R485103
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Contains cells of T. vaginalis, C. albicans, and G. vaginalis for QC of probe systems and direct antigen detection kits.

Thermo Scientific™ TCG-ProbeSwabQC™ contains cells of T. vaginalis, C. albicans, and G. vaginalis for QC of probe systems and direct antigen detection kits.

Stabilized microorganism impregnated swab packaged in individual foil envelope.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Description TCG-ProbeSwabQC™
Format ProbeSwabQC
Product Type Bactidisk
Target Organism Class Organism Sets
Quantity 5 Loops/Pack

The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.