Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ TCH™ Defined Serum Replacement, 50X Concentrate

Catalog No. MP092020026 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP092020026 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP092020026 Supplier MP Biomedicals™ Supplier No. 092020026
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For long-term culturing of cells

  • TCH™ is a fortified, multipurpose serum replacement for long-term culturing of primarily human anchorage-dependent and suspension cells.
  • A universal serum replacement that supports human and mammalian cells; completely free of virus, mycoplasma and other infectious agents; contains no growth factors, steroid hormones, extraneous proteins or phenol red; protein origin - human; Drug Master on file with the FDA.
  • TCH™ Defined Serum Replacement is highly effective in the production of biologicals, as well as in culturing other mammalian cells.
  • This serum replacement is effective in research situations such as studying the effects of growth factors or in the manufacturing of cell derived products.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear, Pink
Content And Storage Store at +4°C.
Form Solution
Product Type Serum Replacement
Quantity 20 mL
Sterility Sterile
Formulation Supplied as a 50X concentrate
For Use With (Application) Highly effective in the production of biologicals, as well as in culturing other mammalian cells.
pH 6.8 to 7.2
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.