Learn More
Aviva Systems Biology Corp TCHP Antibody - C-terminal region (ARP60515_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60515_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TCHP. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human TCHP. Alternate names: MGC10854, TpMsApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-287-070