Learn More
Enzo Life Sciences TCID (10mg). CAS: 30675-13-9
SDP
Supplier:  Enzo Life Sciences BMLEI3990010
Encompass_Preferred
Novel inhibitor of ubiquitin C-terminal hydrolase L3 (IC50 = 0.6µM). Displays 125-fold greater selectivity over UCH-L1 (IC50 = 75µM). Alternative name: 4,5,6,7-Tetrachloroindan-1,3-dione. Purity: ≥95% (TLC). Solubility: Soluble in DMSO (25mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0779