Learn More
TARGETMOL CHEMICALS INC TCID 25MG
SDP
Catalog No. 502579302
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TCID (UCH-L3 Inhibitor)(IC50=0.6 uM) is a DUB inhibitor of ubiquitin C-terminal hydrolase L3. It has the 125-fold selectivity to L1. purity: 99%
Catalog No. 50-257-9302 Supplier TARGETMOL CHEMICALS INC Supplier No. T669725MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TCID (UCH-L3 Inhibitor)(IC50=0.6 uM) is a DUB inhibitor of ubiquitin C-terminal hydrolase L3. It has the 125-fold selectivity to L1. purity: 99%