Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCLP Extraction Fluid 1, pH 4.93 ± 0.05 at 25°C, Ricca Chemical
SDP

Catalog No. 837725 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
10 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
8377-25 10 L
8377-5 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 8377-25 Supplier Ricca Chemical Company Supplier No. 83772.5
Add to Cart
Add to Cart

For use in EPA 1311 Toxicity Characteristic Leaching Procedure

Specifications

Chemical Name or Material TCLP Extraction Fluid 1
Color Colorless
Name Note pH 4.93± 0.05 at 25°C
CAS 1310-73-2, 64-19-7, 7732-18-5
Molecular Weight (g/mol) 40 g/mol
Grade Laboratory
Packaging Cubitainer
pH 4.93
Quantity 10 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.