Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCLP Extraction Fluid 2, pH 2.88 ± 0.05 at 25°C, Ricca Chemical

Click to view available options
Quantity:
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Specifications
Specifications
Chemical Name or Material | TCLP Extraction Fluid 2 |
CAS | 64-19-7, 7732-18-5 |
CAS Min % | 97.0, 0.58 |
CAS Max % | 100.0, 0.61 |
Grade | Laboratory |
Packaging | Cubitainer |
Methods | EPA 1311 Toxicity Characteristic Leaching Procedure |
pH | 2.88 |
Quantity | 20 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction