Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TCLP Semi-Volatile Spiking Mixture, 2000μg/mL in dichloromethane, Ultra Scientific

Catalog No. USTCLP5121
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-TCLP-5121 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-TCLP-5121 Supplier Agilent Technologies Supplier No. TCLP5121
Add to Cart
Add to Cart

Specifications

Chemical Name or Material TCLP Semi-Volatile Spiking Mixture
CAS Max % 99.03, 0.15, 0.15, 0.15, 0.15, 0.15
CAS 95-48-7, 108-39-4, 106-44-5, 106-46-7, 121-14-2
Analyte or Component Names methylene chloride (dichloromethane),o-cresol (2-methylphenol),m-cresol (3-methylphenol)
Element Methylene Chloride, o-cresol, m-cresol (3-methylphenol)
Solvent or Matrix Methylene Chloride
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 2000μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.