Learn More
Medchemexpress LLC Tcmcb07 tfa | 1456699-27-6 | 99.69% | 1344.5 g/mol | 1 MG
SDP
Catalog No. 5000396824
Encompass_Preferred
TCMCB07 TFA is a cyclic nonapeptide, recognized as an orally active and brain-penetrant antagonist of the melanocortin receptor 4 (MC4R). It is noted for its important role in cachexia and exhibits potential transport capabilities.
  • Cyclic nonapeptide MC4R antagonist
  • Orally active and brain-penetrant
  • Important for cachexia research
  • Exhibits potential transport capabilities
  • Known to stimulate food intake and weight gain in rats
Catalog No. 50-003-96824 Supplier Medchemexpress LLC Supplier No. HYP5971A1MG
TCMCB07 TFA is a cyclic nonapeptide, recognized as an orally active and brain-penetrant antagonist of the melanocortin receptor 4 (MC4R). It is noted for its important role in cachexia and exhibits potential transport capabilities.
  • Cyclic nonapeptide MC4R antagonist
  • Orally active and brain-penetrant
  • Important for cachexia research
  • Exhibits potential transport capabilities
  • Known to stimulate food intake and weight gain in rats