Learn More
Medchemexpress LLC Tcmcb07 (TFA) | 99.7% | 1230.46 (free base) | 5 MG
SDP
Catalog No. 5000397827
Encompass_Preferred
TCMCB07 (TFA) is a chemical product from MedChemExpress USA, identified for use as laboratory chemicals and for the manufacture of substances. It is supplied as a solid and is stable under recommended storage conditions.
  • Designed for laboratory use and substance manufacturing
  • Stable under recommended storage conditions
  • Supplied as a solid
  • Storage recommended at -80°C for 2 years or -20°C for 1 year (powder)
  • Sealed storage, away from moisture and light
Catalog No. 50-003-97827 Supplier Medchemexpress LLC Supplier No. HYP5971A5MG
TCMCB07 (TFA) is a chemical product from MedChemExpress USA, identified for use as laboratory chemicals and for the manufacture of substances. It is supplied as a solid and is stable under recommended storage conditions.
  • Designed for laboratory use and substance manufacturing
  • Stable under recommended storage conditions
  • Supplied as a solid
  • Storage recommended at -80°C for 2 years or -20°C for 1 year (powder)
  • Sealed storage, away from moisture and light