Learn More
Medchemexpress LLC Tcmdc-135051 hydrochloride | 2705545-47-5 | 98.2% | 508.05 | C29H34ClN3O3 | 100MG
SDP
Catalog No. 5000239332
Encompass_Preferred
TCMDC-135051 hydrochloride is the hydrochloride salt of TCMDC-135051, a selective and potent inhibitor of Plasmodium falciparum cyclin-dependent-like kinase 3 (PfCLK3) intended for research use in biochemical and cellular studies.
  • selective PfCLK3 inhibition with low off-target toxicity.
  • high purity suitable for research applications.
  • light yellow to yellow solid form for easy handling.
  • stable when stored sealed and protected from moisture.
  • provided in small quantities for assay development and screening.
Catalog No. 50-002-39332 Supplier Medchemexpress LLC Supplier No. HY126323B100MG
TCMDC-135051 hydrochloride is the hydrochloride salt of TCMDC-135051, a selective and potent inhibitor of Plasmodium falciparum cyclin-dependent-like kinase 3 (PfCLK3) intended for research use in biochemical and cellular studies.
  • selective PfCLK3 inhibition with low off-target toxicity.
  • high purity suitable for research applications.
  • light yellow to yellow solid form for easy handling.
  • stable when stored sealed and protected from moisture.
  • provided in small quantities for assay development and screening.