Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TCMDC-135051 hydrochloride | 2705545-47-5 | 98.2% | 508.05 g/mol | C29H34ClN3O3 | 10MM 1ML
SDP

Catalog No. 5000238997
Encompass_Preferred

TCMDC-135051 hydrochloride is a small-molecule inhibitor of the Plasmodium falciparum protein kinase PfCLK3, formulated for in vitro research. It is supplied as a solid and is available as a pre-made 10 mM solution in DMSO for biochemical and antiparasitic assays.

  • High purity (98.17%) suitable for in vitro assays.
  • Molecular weight 508.05 g/mol.
  • Molecular formula C29H34ClN3O3.
  • Soluble in DMSO (≈164 mM); sonication recommended for complete dissolution.
  • Available in multiple solid sizes and as a 10 mM, 1 mL solution in DMSO.
  • Intended for PfCLK3 inhibition and antiparasitic research applications.

Catalog No. 50-002-38997 Supplier Medchemexpress LLC Supplier No. HY126323B10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TCMDC-135051 hydrochloride is a small-molecule inhibitor of the Plasmodium falciparum protein kinase PfCLK3, formulated for in vitro research. It is supplied as a solid and is available as a pre-made 10 mM solution in DMSO for biochemical and antiparasitic assays.

  • High purity (98.17%) suitable for in vitro assays.
  • Molecular weight 508.05 g/mol.
  • Molecular formula C29H34ClN3O3.
  • Soluble in DMSO (≈164 mM); sonication recommended for complete dissolution.
  • Available in multiple solid sizes and as a 10 mM, 1 mL solution in DMSO.
  • Intended for PfCLK3 inhibition and antiparasitic research applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.