Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TCMDC-135051 hydrochloride | 2705545-47-5 | 98.2% | 508.05 g/mol | C29H34ClN3O3 | 5MG
SDP

Catalog No. 5000238354
Encompass_Preferred

TCMDC-135051 hydrochloride is a selective protein kinase PfCLK3 inhibitor with demonstrated antiparasitic activity and includes solubility and stock-preparation guidance for biochemical and cell-based assays.

  • Selective PfCLK3 inhibitor with antiparasitic activity.
  • Antiparasitic activity (EC50 = 320 nM).
  • Hydrochloride salt for improved handling and solubility.
  • Soluble in DMSO (83.33 mg/mL; ultrasonic recommended).
  • Storage guidance for stability in solid and solvent.
  • Light yellow solid appearance.

Catalog No. 50-002-38354 Supplier Medchemexpress LLC Supplier No. HY126323B5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TCMDC-135051 hydrochloride is a selective protein kinase PfCLK3 inhibitor with demonstrated antiparasitic activity and includes solubility and stock-preparation guidance for biochemical and cell-based assays.

  • Selective PfCLK3 inhibitor with antiparasitic activity.
  • Antiparasitic activity (EC50 = 320 nM).
  • Hydrochloride salt for improved handling and solubility.
  • Soluble in DMSO (83.33 mg/mL; ultrasonic recommended).
  • Storage guidance for stability in solid and solvent.
  • Light yellow solid appearance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.