Learn More
Medchemexpress LLC TCN 213 25mg | 556803-08-8 | 25MG
SDP
Catalog No. 5000240320
Encompass_Preferred
TCN 213 is a selective surmountable glycine-dependentlly GluN1/GluN2A NMDAR antagonist with IC50s of 0 55 3 5 40 M in the presence of 75 750 7500 nM glycine respectively TCN 213 can be used to monitor pharmacologically the switch in NMDAR expression in developing cortical neurones[1 [2
Catalog No. 50-002-40320 Supplier Medchemexpress LLC Supplier No. HY10771225MG
TCN 213 is a selective surmountable glycine-dependentlly GluN1/GluN2A NMDAR antagonist with IC50s of 0 55 3 5 40 M in the presence of 75 750 7500 nM glycine respectively TCN 213 can be used to monitor pharmacologically the switch in NMDAR expression in developing cortical neurones[1 [2