Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tcn 213 | 556803-08-8 | MFCD04788042 | 99.2% | 376.54 g/mol | C18H24N4OS2 | 10 MG
SDP

Catalog No. 5000242791
Encompass_Preferred

TCN 213 is a research-grade, glycine-dependent GluN1/GluN2A N-methyl-D-aspartate receptor (NMDAR) antagonist supplied as a solid for biochemical and pharmacological studies. It is intended for use in receptor subtype-selectivity assays and neuropharmacology research.

  • Selective, glycine-dependent GluN1/GluN2A NMDAR antagonist.
  • High purity (99.2%).
  • Chemical formula C18H24N4OS2 and molecular weight 376.54 g/mol.
  • Available in small-milligram quantities for research use.
  • Soluble in DMSO for assay preparation.
  • Store protected from light at refrigerated temperatures; in solution, store at -80°C for long-term stability.

Catalog No. 50-002-42791 Supplier Medchemexpress LLC Supplier No. HY10771210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

TCN 213 is a research-grade, glycine-dependent GluN1/GluN2A N-methyl-D-aspartate receptor (NMDAR) antagonist supplied as a solid for biochemical and pharmacological studies. It is intended for use in receptor subtype-selectivity assays and neuropharmacology research.

  • Selective, glycine-dependent GluN1/GluN2A NMDAR antagonist.
  • High purity (99.2%).
  • Chemical formula C18H24N4OS2 and molecular weight 376.54 g/mol.
  • Available in small-milligram quantities for research use.
  • Soluble in DMSO for assay preparation.
  • Store protected from light at refrigerated temperatures; in solution, store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.