Learn More
Cayman Chemical TCN 237hydrochlorIde 10mg
SDP
Catalog No. 502943484
Encompass_Preferred
An NR2B subunit-containing NMDA receptor antagonist (Ki = 0.85 nM); selectively inhibits glutamate and glycine-induced currents in whole-cell patch-clamp assays using L-M(TK-) cells expressing NR2B subunit-containing NMDA receptors over L-M(TK-) cells expressing NR2A- or NR2D subunit-containing NMDA receptors and HEK293 cells expressing NR2C subunit-containing NMDA receptors; inhibits carrageenan-induced mechanical hyperalgesia in rats at 3 mg/kg
Catalog No. 50-294-3484 Supplier Cayman Chemical Supplier No. 4226610 MG
An NR2B subunit-containing NMDA receptor antagonist (Ki = 0.85 nM); selectively inhibits glutamate and glycine-induced currents in whole-cell patch-clamp assays using L-M(TK-) cells expressing NR2B subunit-containing NMDA receptors over L-M(TK-) cells expressing NR2A- or NR2D subunit-containing NMDA receptors and HEK293 cells expressing NR2C subunit-containing NMDA receptors; inhibits carrageenan-induced mechanical hyperalgesia in rats at 3 mg/kg