Learn More
Medchemexpress LLC HY-14419 5mg Medchemexpress, TCN238 CAS:125404-04-8 Purity:>98%
SDP
Catalog No. 502081599
Encompass_Preferred
Medchemexpress, HY-14419 5mg TCN238 CAS:125404-04-8 TCN238 is an orally bioavailable mGlu4 receptor positive allosteric modulator (PAM) with an EC50 of 1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1599 Supplier Medchemexpress LLC Supplier No. HY144195MG
Medchemexpress, HY-14419 5mg TCN238 CAS:125404-04-8 TCN238 is an orally bioavailable mGlu4 receptor positive allosteric modulator (PAM) with an EC50 of 1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.