Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tco-Amine Hcl Salt

Catalog No. 79045150MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-045-150MG 10 g
79-045-110MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 79-045-150MG Supplier Sigma Aldrich Supplier No. 79045150MG
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Amine functionalized trans-cyclooctene derivative for incorporation of the cyclooctene moiety into carboxyl containing compounds or biomolecules via standard coupling techniques (DCC; EDC etc...). Trans-cyclooctenes are useful in strain-promoted copper-free click chemistry cycloaddition reactions with 1; 2; 4; 5-tetrazines. This cyclooctene will react with tetrazine functionalized compounds or biomolecules without the need for a catalyst to result in a stable covalent linkage. The 4+2 inverse electron demand Diels-Alder cycloaddition between trans-cyclooctene and tetrazines is the fastest biologically compatible ligation technology reported and has had many applications in biological labeling and imaging.

Specifications

Density 1.430 g/mL at 25°C
Molecular Formula C6H6BF9O3
Linear Formula C6H6BF9O3
MDL Number MFCD00093768
Quantity 10 g
Molecular Weight (g/mol) 307.91 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.