Learn More
TARGETMOL CHEMICALS INC TCO-NHS ester 5MG
SDP
Catalog No. 502878561
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TCO-NHS ester is a alkyl/ether-based linker for PROTACs which joins two essential ligands, crucial for forming PROTAC molecules. This linker enables selective protein degradation by leveraging the ubiquitin-proteasome system within cells. Purity 99.77%
Catalog No. 50-287-8561 Supplier TARGETMOL CHEMICALS INC Supplier No. T170145MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TCO-NHS ester is a alkyl/ether-based linker for PROTACs which joins two essential ligands, crucial for forming PROTAC molecules. This linker enables selective protein degradation by leveraging the ubiquitin-proteasome system within cells. Purity 99.77%