Learn More
Medchemexpress LLC Tco-peg-tco | 00-00-0 | 97.1% | 10MG
SDP
Catalog No. 5000262023
Encompass_Preferred
This PEG-based bifunctional linker (average MW 5,000) features trans-cyclooctene (TCO) termini that undergo rapid inverse electron demand Diels-Alder (iEDDA) reactions with tetrazine-functionalized partners. It is designed for assembling bifunctional molecules and rapid bioconjugation under mild conditions, offering solubility and flexibility for use in PROTAC synthesis and other conjugation workflows.
  • Trans-cyclooctene end groups enable fast, selective iEDDA bioorthogonal ligation.
  • Polyethylene glycol backbone provides water solubility and conformational flexibility.
  • Average molecular weight 5,000 gives a defined spacer length for bifunctional constructs.
  • Suitable for PROTAC assembly and diverse bioconjugation workflows.
  • Supplied in small research quantities with batch-specific documentation available.
Catalog No. 50-002-62023 Supplier Medchemexpress LLC Supplier No. HY14073110MG
This PEG-based bifunctional linker (average MW 5,000) features trans-cyclooctene (TCO) termini that undergo rapid inverse electron demand Diels-Alder (iEDDA) reactions with tetrazine-functionalized partners. It is designed for assembling bifunctional molecules and rapid bioconjugation under mild conditions, offering solubility and flexibility for use in PROTAC synthesis and other conjugation workflows.
  • Trans-cyclooctene end groups enable fast, selective iEDDA bioorthogonal ligation.
  • Polyethylene glycol backbone provides water solubility and conformational flexibility.
  • Average molecular weight 5,000 gives a defined spacer length for bifunctional constructs.
  • Suitable for PROTAC assembly and diverse bioconjugation workflows.
  • Supplied in small research quantities with batch-specific documentation available.