Learn More
Medchemexpress LLC TCO-PEG-TCO | 97.1% | 25 MG
SDP
Catalog No. 5000352853
Encompass_Preferred
TCO-PEG-TCO is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, incorporating a TCO group that can engage in an inverse electron demand Diels-Alder reaction (iEDDA) with molecules possessing Tetrazine groups. PROTACs, composed of two distinct ligands connected by a linker, target an E3 ubiquitin ligase and a specific protein, leveraging the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Incorporates a TCO group
  • Engages in inverse electron demand Diels-Alder reaction (iEDDA)
  • Leverages the intracellular ubiquitin-proteasome system
  • For selective degradation of target proteins
Catalog No. 50-003-52853 Supplier Medchemexpress LLC Supplier No. 25MGHY140731
TCO-PEG-TCO is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, incorporating a TCO group that can engage in an inverse electron demand Diels-Alder reaction (iEDDA) with molecules possessing Tetrazine groups. PROTACs, composed of two distinct ligands connected by a linker, target an E3 ubiquitin ligase and a specific protein, leveraging the intracellular ubiquitin-proteasome system for selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Incorporates a TCO group
  • Engages in inverse electron demand Diels-Alder reaction (iEDDA)
  • Leverages the intracellular ubiquitin-proteasome system
  • For selective degradation of target proteins