Learn More
Medchemexpress LLC TCO-PEG-TCO (MW 5000) | 97.1% | 100 MG
SDP
Catalog No. 5000355647
Encompass_Preferred
TCO-PEG-TCO (MW 5000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This click chemistry reagent contains a TCO group, enabling it to undergo an inverse electron demand Diels-Alder reaction (iEDDA) with molecules that contain Tetrazine groups.
  • PEG-based PROTAC linker for targeted protein degradation.
  • Facilitates click chemistry reactions with Tetrazine groups.
  • Utilizes the intracellular ubiquitin-proteasome system.
Catalog No. 50-003-55647 Supplier Medchemexpress LLC Supplier No. HY140731100MG
TCO-PEG-TCO (MW 5000) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. This click chemistry reagent contains a TCO group, enabling it to undergo an inverse electron demand Diels-Alder reaction (iEDDA) with molecules that contain Tetrazine groups.
  • PEG-based PROTAC linker for targeted protein degradation.
  • Facilitates click chemistry reactions with Tetrazine groups.
  • Utilizes the intracellular ubiquitin-proteasome system.