Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TCO-PEG-TCO (MW 5000) | 97.1% | 50 MG
SDP

Catalog No. 5000356653
Encompass_Preferred

TCO-PEG-TCO (MW 5000) is a PEG-based PROTAC linker, a click chemistry reagent containing a TCO group that undergoes an inverse electron demand Diels-Alder reaction (iEDDA) with Tetrazine groups. It is utilized in the synthesis of PROTACs, which consist of two ligands connected by a linker-one targeting an E3 ubiquitin ligase and the other a protein of interest. These compounds leverage the intracellular ubiquitin-proteasome system to facilitate selective degradation of target proteins. This product has an average molecular weight of 5000 and appears as a white to off-white solid.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains a TCO group for iEDDA reaction with tetrazine groups
  • Facilitates selective degradation of target proteins
  • Molecular weight: 5000 (average)
  • Appearance: Solid
  • Color: White to off-white
  • For research use only

Catalog No. 50-003-56653 Supplier Medchemexpress LLC Supplier No. HY14073150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TCO-PEG-TCO (MW 5000) is a PEG-based PROTAC linker, a click chemistry reagent containing a TCO group that undergoes an inverse electron demand Diels-Alder reaction (iEDDA) with Tetrazine groups. It is utilized in the synthesis of PROTACs, which consist of two ligands connected by a linker-one targeting an E3 ubiquitin ligase and the other a protein of interest. These compounds leverage the intracellular ubiquitin-proteasome system to facilitate selective degradation of target proteins. This product has an average molecular weight of 5000 and appears as a white to off-white solid.

  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains a TCO group for iEDDA reaction with tetrazine groups
  • Facilitates selective degradation of target proteins
  • Molecular weight: 5000 (average)
  • Appearance: Solid
  • Color: White to off-white
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.