Learn More
Broadpharm TCO-PEG12-NHS ester, 2185016-39-9, MFCD30536161, 10mg
Catalog No. NC1690663
Encompass
Formula: C40H70N2O18, Purity: 98%, MW: 867.0, TCO-PEG12-NHS Ester is a PEG derivative containing a TCO moiety and a NHS ester. The hydrophilic PEG spacer increases water-solubility and a decreases steric hindrance during ligation. This reagent can be used to label antibodies, proteins and other primary amine-containing macromolecules with TCO moiety.
Catalog No. NC1690663 Supplier Broadpharm Supplier No. BP24123
Formula: C40H70N2O18, Purity: 98%, MW: 867.0, TCO-PEG12-NHS Ester is a PEG derivative containing a TCO moiety and a NHS ester. The hydrophilic PEG spacer increases water-solubility and a decreases steric hindrance during ligation. This reagent can be used to label antibodies, proteins and other primary amine-containing macromolecules with TCO moiety.