Learn More
Medchemexpress LLC TCO-PEG2-amine | 2141981-87-3 | 97.0% | C15H28N2O4 | 100 MG
SDP
Catalog No. 5000355077
Encompass_Preferred
TCO-PEG2-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. These molecules consist of two distinct ligands connected by a linker, where one targets an E3 ubiquitin ligase and the other targets the protein of interest. PROTACs function by harnessing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only
Catalog No. 50-003-55077 Supplier Medchemexpress LLC Supplier No. HY141177100MG
TCO-PEG2-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. These molecules consist of two distinct ligands connected by a linker, where one targets an E3 ubiquitin ligase and the other targets the protein of interest. PROTACs function by harnessing the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • For research use only