Learn More
Medchemexpress LLC TCO-PEG2-amine | 2141981-87-3 | ≥97.0% | C15H28N2O4 | 25 MG
SDP
Catalog No. 5000357263
Encompass_Preferred
TCO-PEG2-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed to degrade target proteins by connecting an E3 ubiquitin ligase ligand to a target protein ligand via a linker, leveraging the intracellular ubiquitin-proteasome system. This product appears as a colorless to light yellow liquid.
  • PEG-based PROTAC linker
  • Suitable for the synthesis of PROTACs
  • Purity ≥97.0%
  • Colorless to light yellow liquid appearance
  • For research use only
Catalog No. 50-003-57263 Supplier Medchemexpress LLC Supplier No. 25MGHY141177
TCO-PEG2-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. PROTACs are designed to degrade target proteins by connecting an E3 ubiquitin ligase ligand to a target protein ligand via a linker, leveraging the intracellular ubiquitin-proteasome system. This product appears as a colorless to light yellow liquid.
  • PEG-based PROTAC linker
  • Suitable for the synthesis of PROTACs
  • Purity ≥97.0%
  • Colorless to light yellow liquid appearance
  • For research use only