Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TCO-PEG2-amine | 2141981-87-3 | ≥97.0% | C15H28N2O4 | 50 MG
SDP

Catalog No. 5000354901
Encompass_Preferred

TCO-PEG2-amine is a PEG-based PROTAC linker. This compound is used in the synthesis of PROTACs.

  • Purity of ≥97.0%
  • Molecular formula C15H28N2O4
  • Molecular weight 300.39
  • Appears as a colorless to light yellow liquid
  • Recommended storage at 4°C, protected from light
  • Suitable for PROTAC synthesis

Catalog No. 50-003-54901 Supplier Medchemexpress LLC Supplier No. HY14117750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TCO-PEG2-amine is a PEG-based PROTAC linker. This compound is used in the synthesis of PROTACs.

  • Purity of ≥97.0%
  • Molecular formula C15H28N2O4
  • Molecular weight 300.39
  • Appears as a colorless to light yellow liquid
  • Recommended storage at 4°C, protected from light
  • Suitable for PROTAC synthesis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.