Learn More
Medchemexpress LLC TCO-PEG3-alcohol | 1955497-82-1 | 99.3% | C17H31NO6 | 10MG
SDP
Catalog No. 5000259058
Encompass_Preferred
TCO-PEG3-alcohol is a PEG-based linker and bioorthogonal click chemistry reagent featuring a trans-cyclooctene (TCO) group and a terminal alcohol. It undergoes rapid inverse electron-demand Diels-Alder (iEDDA) reactions with tetrazine-functionalized partners, making it useful for PROTAC assembly, bioconjugation, and selective labeling in chemical biology. High purity and defined molecular weight support synthetic applications.
  • Enables rapid iEDDA click reactions with tetrazines.
  • Provides a PEG3 spacer for improved solubility and flexibility.
  • Contains a terminal alcohol for downstream functionalization.
  • High purity suitable for synthesis and conjugation workflows.
  • Compatible with PROTAC synthesis and bioconjugation applications.
Catalog No. 50-002-59058 Supplier Medchemexpress LLC Supplier No. HY13347610MG
TCO-PEG3-alcohol is a PEG-based linker and bioorthogonal click chemistry reagent featuring a trans-cyclooctene (TCO) group and a terminal alcohol. It undergoes rapid inverse electron-demand Diels-Alder (iEDDA) reactions with tetrazine-functionalized partners, making it useful for PROTAC assembly, bioconjugation, and selective labeling in chemical biology. High purity and defined molecular weight support synthetic applications.
  • Enables rapid iEDDA click reactions with tetrazines.
  • Provides a PEG3 spacer for improved solubility and flexibility.
  • Contains a terminal alcohol for downstream functionalization.
  • High purity suitable for synthesis and conjugation workflows.
  • Compatible with PROTAC synthesis and bioconjugation applications.