Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tco-Peg3-Maleimide

Catalog No. 7904435MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-044-35MG 10 mg
79-044-325MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 79-044-35MG Supplier Sigma Aldrich Supplier No. 7904435MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Maleimide (thiol reactive) functionalized trans-cyclooctene derivative for incorporation of the cyclooctene moiety into thiol containing compounds or biomolecules. Trans-cyclooctenes are useful in strain-promoted copper-free click chemistry cycloaddition

Specifications

Molecular Formula C12H22N2O2 -+ HCl
Linear Formula C12H22N2O2 -+ HCl
Quantity 10 mg
Synonym trans-Cyclooctene-amine hydrochloride salt; TCO-NH2, HCl salt
Molecular Weight (g/mol) 262.78 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.