Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vector Laboratories VECTOR LABORATORIES LLC TCO-PEG3-Maleimide, MW 523.62, Purity >95%
SDP

Catalog No. NC3973230
Encompass

TCO-PEG3-Maleimide enables simple and efficient incorporation of TCO moiety onto antibodies, cysteine-containing peptides and other thiol-containing molecules. The maleimide group specifically and efficiently reacts with reduced thiols (sulfhydryl groups, –SH) at pH 6.5 to 7.5 to form stable thioether bonds. The hydrophilic polyethylene glycol (PEG) spacer arm improves solubility in aqueous buffers.

  • CAS number: 1609659-01-9
  • Molecular weight: 523.62
  • Molecular formula: C26H41N3O8
  • Appearance: Colorless to slightly yellow oil
  • Purity: >95% (HPLC)
  • Solubility: DMSO, DMF, DCM, THF, Chloroform
  • Storage instructions: −50°C to −85°C
  • Shipping conditions: Dry ice
  • For research use only.

Catalog No. NC3973230 Supplier Vector Laboratories Supplier No. CCT10021
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TCO-PEG3-Maleimide enables simple and efficient incorporation of TCO moiety onto antibodies, cysteine-containing peptides and other thiol-containing molecules. The maleimide group specifically and efficiently reacts with reduced thiols (sulfhydryl groups, –SH) at pH 6.5 to 7.5 to form stable thioether bonds. The hydrophilic polyethylene glycol (PEG) spacer arm improves solubility in aqueous buffers.

  • CAS number: 1609659-01-9
  • Molecular weight: 523.62
  • Molecular formula: C26H41N3O8
  • Appearance: Colorless to slightly yellow oil
  • Purity: >95% (HPLC)
  • Solubility: DMSO, DMF, DCM, THF, Chloroform
  • Storage instructions: −50°C to −85°C
  • Shipping conditions: Dry ice
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.