Learn More
Medchemexpress LLC TCO-PEG4-NHS ester | 1621096-79-4 | 99.2% | C24H38N2O10 | 25 MG
SDP
Catalog No. 5000352785
Encompass_Preferred
TCO-PEG4-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring a TCO group that facilitates an inverse electron demand Diels-Alder reaction (iEDDA) with molecules containing Tetrazine groups. PROTACs are composed of two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • TCO group facilitates inverse electron demand Diels-Alder reaction (iEDDA) with Tetrazine groups
  • Utilizes intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • For research use only
Catalog No. 50-003-52785 Supplier Medchemexpress LLC Supplier No. 25MGHY141167A
TCO-PEG4-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. This compound functions as a click chemistry reagent, featuring a TCO group that facilitates an inverse electron demand Diels-Alder reaction (iEDDA) with molecules containing Tetrazine groups. PROTACs are composed of two distinct ligands connected by a linker: one ligand targets an E3 ubiquitin ligase, and the other targets a specific protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • TCO group facilitates inverse electron demand Diels-Alder reaction (iEDDA) with Tetrazine groups
  • Utilizes intracellular ubiquitin-proteasome system to selectively degrade target proteins
  • For research use only