Learn More
Medchemexpress LLC TCO-PEG4-NHS ester | 1621096-79-4 | 99.2% | C24H38N2O10 | 50 MG
SDP
Catalog No. 5000355627
Encompass_Preferred
TCO-PEG4-NHS ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It also functions as a click chemistry reagent, capable of undergoing an inverse electron demand Diels-Alder reaction (iEDDA) with molecules containing Tetrazine groups. This reagent facilitates the creation of PROTACs, which leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains a TCO group
  • Undergoes inverse electron demand Diels-Alder reaction (iEDDA) with Tetrazine groups
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55627 Supplier Medchemexpress LLC Supplier No. HY141167A50MG
TCO-PEG4-NHS ester is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It also functions as a click chemistry reagent, capable of undergoing an inverse electron demand Diels-Alder reaction (iEDDA) with molecules containing Tetrazine groups. This reagent facilitates the creation of PROTACs, which leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains a TCO group
  • Undergoes inverse electron demand Diels-Alder reaction (iEDDA) with Tetrazine groups
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins