Learn More
Broadpharm TCO-PEG4-NHS ester, 1613439-69-2, MFCD28118912, 25mg
Catalog No. NC1622814
Encompass
Formula: C24H38N2O10, Purity: 95%, MW: 514.6, TCO-PEG4-NHS Ester is a PEG derivative containing a TCO moiety and a NHS ester. The hydrophilic PEG spacer increases water-solubility and a decreases steric hindrance during ligation. This reagent can be used to label antibodies, proteins and other primary amine-containing macromolecules with TCO moiety.
Catalog No. NC1622814 Supplier Broadpharm Supplier No. BP2241825MG
Formula: C24H38N2O10, Purity: 95%, MW: 514.6, TCO-PEG4-NHS Ester is a PEG derivative containing a TCO moiety and a NHS ester. The hydrophilic PEG spacer increases water-solubility and a decreases steric hindrance during ligation. This reagent can be used to label antibodies, proteins and other primary amine-containing macromolecules with TCO moiety.