Learn More
TARGETMOL CHEMICALS INC TCO-PEG4-NHS ester 25MG
SDP
Catalog No. 502878478
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TCO-PEG4-NHS ester is a PROTAC linker bridge in the PEG class, used for synthesizing various PROTAC molecules. As a cleavable and synthetic ADC, it is also utilized in the synthesis of antibody-drug conjugates (ADCs).
Catalog No. 50-287-8478 Supplier TARGETMOL CHEMICALS INC Supplier No. T1701725MG
Also available in 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. TCO-PEG4-NHS ester is a PROTAC linker bridge in the PEG class, used for synthesizing various PROTAC molecules. As a cleavable and synthetic ADC, it is also utilized in the synthesis of antibody-drug conjugates (ADCs).