Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tco Peg4 Succinimidyl Ester

Catalog No. 7904355MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-043-55MG 5 mg
79-043-525MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 79-043-55MG Supplier Sigma Aldrich Supplier No. 7904355MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Succinimidyl ester/NHS (amine reactive) functionalized trans-cyclooctene derivative for incorporation of the cyclooctene moiety into amine containing compounds or biomolecules. Trans-cyclooctenes are useful in strain-promoted, copper-free, click chemistry cycloaddition reactions with 1, 2, 4, 5-tetrazines. This cyclooctene will react with tetrazine functionalized compounds or biomolecules, without the need for a catalyst, to result in a stable covalent linkage. The 4+2 inverse electron demand Diels-Alder cycloaddition between trans-cyclooctene and tetrazines is the fastest biologically compatible ligation technology reported, and has had many applications in biological labeling and imaging. The PEG spacer allows for increased water solubility, less aggregation, and an increased distance between the amine to be modified and the reactive alkene.

Specifications

Molecular Formula C24H38N2O10
Linear Formula C24H38N2O10
MDL Number MFCD28118912
Quantity 5 mg
Synonym trans-Cyclooctene-PEG4-NHS
Molecular Weight (g/mol) 514.57 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.