Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TCO-PEG4-VC-PAB-MMAE | 2758671-45-1 | 99.3% | 1522.91 | C78H127N11O19 | 10 MG
SDP

Catalog No. 5000203404
Encompass_Preferred

TCO-PEG4-VC-PAB-MMAE is a drug-linker conjugate intended for use in antibody-drug conjugate (ADC) assembly and click-chemistry bioconjugation workflows. It combines a trans-cyclooctene (TCO) click handle, a PEG4-Val-Cit-PAB cleavable linker, and the cytotoxic payload MMAE to enable site-selective conjugation and intracellular release of the payload.

  • Provides a trans-cyclooctene (TCO) handle for inverse electron-demand Diels-Alder reactions with tetrazines.
  • Contains a PEG4-Val-Cit-PAB cleavable linker for enzymatic payload release.
  • Conjugated to MMAE to supply a potent cytotoxic payload for ADC research.
  • High purity (99.3%) suitable for research and bioconjugation workflows.
  • Characterized by molecular weight 1522.91 and formula C78H127N11O19.

Catalog No. 50-002-03404 Supplier Medchemexpress LLC Supplier No. HY14805710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TCO-PEG4-VC-PAB-MMAE is a drug-linker conjugate intended for use in antibody-drug conjugate (ADC) assembly and click-chemistry bioconjugation workflows. It combines a trans-cyclooctene (TCO) click handle, a PEG4-Val-Cit-PAB cleavable linker, and the cytotoxic payload MMAE to enable site-selective conjugation and intracellular release of the payload.

  • Provides a trans-cyclooctene (TCO) handle for inverse electron-demand Diels-Alder reactions with tetrazines.
  • Contains a PEG4-Val-Cit-PAB cleavable linker for enzymatic payload release.
  • Conjugated to MMAE to supply a potent cytotoxic payload for ADC research.
  • High purity (99.3%) suitable for research and bioconjugation workflows.
  • Characterized by molecular weight 1522.91 and formula C78H127N11O19.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.