Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC TCO-PEG4-VC-PAB-MMAE | 2758671-45-1 | 99.3% | 1522.91 g/mol | C78H127N11O19 | 5 MG
SDP

Catalog No. 5000202114
Encompass_Preferred

TCO-PEG4-VC-PAB-MMAE is an antibody-drug conjugate (ADC) drug-linker conjugate that combines a cleavable valine-citrulline-para-aminobenzyl (VC-PAB) linker with the cytotoxic payload monomethyl auristatin E (MMAE). It contains a trans-cyclooctene (TCO) group for inverse-electron-demand Diels-Alder (iEDDA) click conjugation to tetrazine-functionalized molecules and is supplied as a solid for ADC research and bioconjugation workflows.

  • Contains TCO group for iEDDA click chemistry.
  • Cleavable valine-citrulline-PAB linker enabling controlled payload release.
  • MMAE payload with potent tubulin inhibitory activity.
  • High purity suitable for research applications.
  • Storage: solid at -20°C; protect from light.

Catalog No. 50-002-02114 Supplier Medchemexpress LLC Supplier No. HY1480575MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TCO-PEG4-VC-PAB-MMAE is an antibody-drug conjugate (ADC) drug-linker conjugate that combines a cleavable valine-citrulline-para-aminobenzyl (VC-PAB) linker with the cytotoxic payload monomethyl auristatin E (MMAE). It contains a trans-cyclooctene (TCO) group for inverse-electron-demand Diels-Alder (iEDDA) click conjugation to tetrazine-functionalized molecules and is supplied as a solid for ADC research and bioconjugation workflows.

  • Contains TCO group for iEDDA click chemistry.
  • Cleavable valine-citrulline-PAB linker enabling controlled payload release.
  • MMAE payload with potent tubulin inhibitory activity.
  • High purity suitable for research applications.
  • Storage: solid at -20°C; protect from light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.