Learn More
Medchemexpress LLC TCO-PEG5-C2-Mal | 2748951-17-7 | 95.0% | 512.59 | 5 MG
SDP
Catalog No. 5000394257
Encompass_Preferred
TCO-PEG5-C2-Mal is a heterobifunctional linker that can be used for tetrazine ligation and sulfhydryl conjugation.
Catalog No. 50-003-94257 Supplier Medchemexpress LLC Supplier No. HY1647855MG
TCO-PEG5-C2-Mal is a heterobifunctional linker that can be used for tetrazine ligation and sulfhydryl conjugation.