Learn More
Medchemexpress LLC Tco-peg8-nhs ester | 2353409-95-5 | 95.0% | C32H54N2O14 | 10MG
SDP
Catalog No. 5000261396
Encompass_Preferred
TCO-PEG8-NHS ester is a PEG-based PROTAC linker with an N-hydroxysuccinimide (NHS) activated ester that enables efficient conjugation to primary amines. The molecule incorporates a trans-cyclooctene (TCO) moiety separated by an eight-unit PEG spacer to provide aqueous solubility and flexible spacing for bioconjugation and targeted protein degradation construct synthesis.
  • NHS-activated ester for efficient amide bond formation.
  • Eight-unit PEG spacer improves solubility and flexibility.
  • Provides a trans-cyclooctene (TCO) handle for click chemistry.
  • High reported purity (95.0%).
  • Supplied in small-mass packages suitable for research use.
Catalog No. 50-002-61396 Supplier Medchemexpress LLC Supplier No. HY14116910MG
TCO-PEG8-NHS ester is a PEG-based PROTAC linker with an N-hydroxysuccinimide (NHS) activated ester that enables efficient conjugation to primary amines. The molecule incorporates a trans-cyclooctene (TCO) moiety separated by an eight-unit PEG spacer to provide aqueous solubility and flexible spacing for bioconjugation and targeted protein degradation construct synthesis.
  • NHS-activated ester for efficient amide bond formation.
  • Eight-unit PEG spacer improves solubility and flexibility.
  • Provides a trans-cyclooctene (TCO) handle for click chemistry.
  • High reported purity (95.0%).
  • Supplied in small-mass packages suitable for research use.