Learn More
Medchemexpress LLC TCO-PEG8-NHS ester | 2353409-95-5 | 95.0% | C32H54N2O14 | 25 MG
SDP
Catalog No. 5000352492
Encompass_Preferred
TCO-PEG8-NHS ester is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs, which are compounds containing two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for a target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase
  • Other ligand for a target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-52492 Supplier Medchemexpress LLC Supplier No. 25MGHY141169
TCO-PEG8-NHS ester is a PEG-based PROTAC linker. It can be used in the synthesis of PROTACs, which are compounds containing two different ligands connected by a linker: one for an E3 ubiquitin ligase and the other for a target protein. PROTACs utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • One ligand for an E3 ubiquitin ligase
  • Other ligand for a target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins